Learn More
Letrazuril, MedChemExpress
Product Code. 16465066
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16465066
Brand: Medchem Express HY106859
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Letrazuril is an anti-HIV agent.
Letrazuril is an anti-HIV agent.
Specifications
Chemical Name or Material Letrazuril
CAS 103337-74-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H9Cl2FN4O2
Quantity 100 mg
Solubility Information DMSO : ≥ 165 mg/mL (421.80 mM)
SMILES O=C(N1)N(N=CC1=O)C2=CC(Cl)=C(C(C3=CC=C(F)C=C3)C#N)C(Cl)=C2
Molecular Weight (g/mol) 391.18
Formula Weight 391.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus