Learn More
Leonurine, TRC
Specifications
Specifications
| Chemical Name or Material | Leonurine |
| CAS | 24697-74-3 |
| Molecular Formula | C14H21N3O5 |
| Quantity | 100 mg |
| Synonym | 4-Guanidino-1-butanol Syringate, 4-Hydroxy-3,5-dimethoxybenzoic Acid, Δ-guanidinobutyl Ester, Leonurin, SCM 198, Syringic Acid Δ-guanidinobutyl Ester, [4-(4-Hydroxy-3,5-dimethoxybenzoyloxy)butyl]guanidine, 4-Guanidinobutyl 4-hydroxy-3,5-dimethoxybenzoate |
| InChI Formula | InChI=1S/C14H21N3O5/c1-20-10-7-9(8-11(21-2)12(10)18)13(19)22-6-4-3-5-17-14(15)16/h7-8,18H,3-6H2,1-2H3,(H4,15,16,17) |
| SMILES | CN(C)CCc1c2c3c4c(c(OC)ccc4C(=O)Oc3c(OC)c1)OC2=O |
| Molecular Weight (g/mol) | 311.33 |