Learn More
Emma Safety Footwear Leo All-Round Safety D-Fit Shoes
Click to view available options
Size:
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Description
- Full grain leather uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
- Strobel-stitched upper construction
Specifications
Specifications
| Color | Black |
| Liner Material | Hydro-Tec Sanitized Silver lining |
| Material | Leather |
| Size | 40 |
| Sole Features | Non-woven antistatic insole |
| Sole Material | GripForce™ Easy Twist outsole |
| Toe Material | Steel |
| Product Type | All-Round Safety Shoes |