Learn More
3M™ Super-Clear™ Lens Cleaning Tissue Station
Ideal for keeping safety and prescription spectacles clear. 3M™ Super-Clear™ Lens Cleaning Tissue Station comes ready to hang on the wall.
Brand: 3M™ 7000062429
Additional Details : Weight : 0.01000kg
Description
- Anti-static and anti-fog
- For CR39, polycarbonate and glass lenses
- Contains 473mL pump and bottle of cleaning fluid
- Contains 1500 sheets of disposable, pre-moistened lens cleaning paper