missing translation for 'onlineSavingsMsg'
Learn More

Lenalidomide, MedChemExpress

Product Code. 16441966
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Pakningsstørrelse:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16441966 500 mg 500mg
16457976 100 mg 100mg
16441999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16441966 Leverandør Medchem Express Leverandørnr. HYA0003

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Lenalidomide (CC-5013), a derivative of Thalidomide, acts as molecular glue.

Lenalidomide is an orally active immunomodulator. Lenalidomide (CC-5013) is a ligand of ubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide (CC-5013) specifically inhibits growth of mature B-cell lymphomas, including multiple myeloma, and induces IL-2 release from T cells.

Tekniske data

Chemical Name or Material Lenalidomide
CAS 191732-72-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H13N3O3
Quantity 500 mg
Synonym CC-5013
Solubility Information DMSO : 100 mg/mL (385.71 mM; Need ultrasonic)
SMILES O=C1C2=CC=CC(N)=C2CN1C(C(N3)=O)CCC3=O
Molecular Weight (g/mol) 259.26
Formula Weight 259.26
Percent Purity 99.05%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.