Learn More
Tocris Bioscience™ Lenalidomide 4'-PEG2-amine
Description
Lenalidomide 4'-PEG2-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 3-[4-[[2-[2-(2-(2-Aminoethoxy)ethoxy]ethyl]amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione dihydrochloride |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C19H26N4O50.2 HCl |
| Purity | 0.95 |