missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legionella Antibody (LP46), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64479CL1
This item is not returnable.
View return policy
Description
Legionella Monoclonal antibody specifically detects Legionella in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| Legionella | |
| Monoclonal | |
| CoraFluor 1 | |
| L. pneumophila, Legionellosis | |
| Native Legionella | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG3 |
| Immunocytochemistry/Immunofluorescence | |
| LP46 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction