Learn More
Medline Scientific™ LED Ring Illuminator
Product Code. p-7138533 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Steddy Stereo Model Microscopes
Varizoom Stereo Model Microscopes
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 12653035
Brand: Medline Scientific™ 7062.5001
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Used to provide flat uniform lighting of a subject during stereo microscopy.
  • With separate power supply and light intensity control
  • For stereo models only
Specifications
Type Ring Illuminator
Description LED
For Use With (Equipment) Varizoom Stereo Model Microscopes
Includes Separate power source and light intensity control
Wattage 9 w