missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Leaded Brass, CZ115, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Leaded Brass, CZ115, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Chromium/Aluminium/Arsenic/Antimony/Silver/Phosphorus/Nickel/Iron/Tin/Manganese/Lead/Zinc/Copper/Silicon |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Chromium 0.0005 %/Aluminium 0.0007 %/Arsenic 0.0008 %/Antimony 0.002 %/Silver 0.0041 %/Phosphorus 0.0091 %/Nickel 0.0143 %/Iron 0.601 %/Tin 0.729 %/Manganese 1.095 %/Lead 1.169 %/Zinc 39.2 %/Copper 57.19 %/Silicon (<0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction