missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Pb/Sn/Ca Battery Alloy, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Lead, Pb/Sn/Ca Battery Alloy, 50g
Specifications
Specifications
| Chemical Name or Material | Arsenic/Copper/Cadmium/Antimony/Zinc/Silver/Bismuth/Aluminium/Calcium/Tin |
| CAS | 7439-92-1 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.00053 %/Copper 0.0011 %/Cadmium 0.0013 %/Antimony 0.0047 %/Zinc 0.0062 %/Silver 0.0095 %/Bismuth 0.014 %/Aluminium 0.0161 %/Calcium 0.0865 %/Tin 0.941 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction