missing translation for 'onlineSavingsMsg'
Learn More

LDN-212320, MedChemExpress

Product Code. 16411517
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16411517

Brand: Medchem Express HY12741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LDN-212320 (LDN-0212320) is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator (at translational level). LDN-212320 (LDN-0212320) prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC[1].

LDN-212320 (LDN-0212320) is a glutamate transporter (GLT-1)/excitatory amino acid transporter 2 (EAAT2) activator (at translational level). LDN-212320 (LDN-0212320) prevents nociceptive pain by upregulating astroglial GLT-1 expression in the hippocampus and ACC[1].

Specifications

Chemical Name or Material LDN-212320
Physical Form Solid
CAS 894002-50-7
Color Gray
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H15N3S
For Use With (Application) Neuroscience-Neuromodulation
Synonym LDN-0212320 OSU-0212320
Solubility Information DMSO : 50 mg/mL (170.42 mM; Need ultrasonic)
SMILES CC1=CC=CC=C1CSC2=NN=C(C3=NC=CC=C3)C=C2
Molecular Weight (g/mol) 293.39
Formula Weight 293.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.