missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LDLRAD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15613985
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15613985 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613985 Supplier Invitrogen™ Supplier No. PA550050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total LDLRAD2 protein.

The exact functions of this protein remain unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLRAD2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LDLRAD2
Gene Accession No. Q5SZI1
Gene Alias LDLRAD2; low density lipoprotein receptor A domain containing 2; low density lipoprotein receptor class A domain containing 2; low-density lipoprotein receptor class A domain-containing protein 2; lrad2
Gene Symbols LDLRAD2
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human LDLRAD2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401944, 435811
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.