missing translation for 'onlineSavingsMsg'
Learn More

LDC7559, MedChemExpress

Product Code. p-200095892
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487227

Brand: Medchem Express HY111674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LDC7559 is a gasdermin D (GSDMD) inhibitor via blocking neutrophil extracellular trap (NET) in the late stages .

LDC7559 is a gasdermin D (GSDMD) inhibitor via blocking neutrophil extracellular trap (NET) in the late stages .

Specifications

Chemical Name or Material LDC7559
CAS 2407782-01-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H19N3O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (286.22 mM; Need ultrasonic)
SMILES COC1=C(C=CC=C1)C2=NN3C(COC(C=C(NC(C)=O)C=C4)=C4C3)=C2
Molecular Weight (g/mol) 349.38
Formula Weight 349.38
Percent Purity 99.15%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.