Learn More
LDC7559, MedChemExpress
Description
LDC7559 is a gasdermin D (GSDMD) inhibitor via blocking neutrophil extracellular trap (NET) in the late stages .
Specifications
Specifications
| Chemical Name or Material | LDC7559 |
| CAS | 2407782-01-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H19N3O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (286.22 mM; Need ultrasonic) |
| SMILES | COC1=C(C=CC=C1)C2=NN3C(COC(C=C(NC(C)=O)C=C4)=C4C3)=C2 |
| Molecular Weight (g/mol) | 349.38 |
| Formula Weight | 349.38 |
| Show More |