missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCORL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17468-1-AP-150UL
This item is not returnable.
View return policy
Description
May act as transcription activator that binds DNA elements with the sequence 5′-CCCTATCGATCGATCTCTACCT-3′. May play a role in spermatogenesis.Specifications
| LCORL | |
| Polyclonal | |
| Unconjugated | |
| LCORL | |
| LCoR like protein, LCORL, MLR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 209707, 254251, 498385 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3U285, Q8N3X6 | |
| Lcorl | |
| LCORL Fusion Protein Ag11377 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction