missing translation for 'onlineSavingsMsg'
Learn More

Lck inhibitor 2, MedChemExpress

Product Code. 16454088
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454088

Brand: Medchem Express HY10644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lck inhibitor 2 is a bis-anilinopyrimidine inhibitor of tyrosine kinases including LCK, BTK, LYN, SYK, and TXK. The IC50 values are 13nM, 9nM, 3nM, 26nM and 2nM for Lck, Btk, Lyn, Btk and Txk respectively

Lck inhibitor 2 is a bis-anilinopyrimidine inhibitor of tyrosine kinases including LCK, BTK, LYN, SYK, and TXK. The IC50 values are 13nM, 9nM, 3nM, 26nM and 2nM for Lck, Btk, Lyn, Btk and Txk respectively

Specifications

Chemical Name or Material Lck inhibitor 2
CAS 944795-06-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H17N5O2
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (59.64 mM; Need ultrasonic)
SMILES OC1=CC=C(C)C(NC2=CC=NC(NC3=CC(C(N)=O)=CC=C3)=N2)=C1
Molecular Weight (g/mol) 335.36
Formula Weight 335.36
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.