missing translation for 'onlineSavingsMsg'
Learn More

Hach™ LCK Cuvette Tests

Product Code. 10376144
Change view
Click to view available options
Analytes:
Acid Capacity (KS4.3/CaCO3)
Dissolved Sulphide
Iron II and III
Unit Size:
Each
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analytes Unit Size
10376144 Iron II and III Each
10122744 Acid Capacity (KS4.3/CaCO3) Each
10182404 Dissolved Sulphide Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376144 Supplier Hach™ Supplier No. LCK320
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    146.48 € / Pack of 1
    Save 4.52 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Insert, read and finish. Hach™ LCK Cuvette Tests are pre-dosed reagents packaged in a fully equipped box. They provide maximum safety, ease of use and photometric evaluation, while using a minimum amount of reagent/chemicals for an environmentally friendly solution.

Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.

  • Pre-dosed, ready-to-use reagents with no rinsing steps needed
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding

Order Info

dimRange

TRUSTED_SUSTAINABILITY

Specifications

Analytes Iron II and III
Certifications/Compliance DIN 38405-D17
Color Red
Format Pre-dosed Tubes
For Use With (Application) Water quality testing
For Use With (Equipment) Photometer (DR3900, DR6000, DR1900, DR2800, DR3800, DR5000), Addista standard solutions
Includes Fully equipped box, reagent set
No. of Tests 24
Range 0.2 to 6.0mg/L Fe
Storage Requirements 2°C to 8°C
Test Method 1.10-Phenanthroline
Type Iron (II/III) Cuvette Test
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron (II/III), cuvette test Measuring range 0.2-6. 0 mg/l
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: citric acid, polyvidon 25, D-mannitol, 1,10-phenanthroline chloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.