Learn More
(±)-Lavandulyl Acetate, TRC
Product Code. 30009798
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009798 100 mg 100mg
30012747 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009798 Supplier Toronto Research Chemicals Supplier No. TRCL200000100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25905-14-0
Molecular Formula C12 H20 O2
Molecular Weight (g/mol) 196.29
Synonym 4-Hexen-1-ol, 5-methyl-2-(1-methylethenyl)-, 1-acetate, 4-Hexen-1-ol, 2-isopropenyl-5-methyl-, acetate (8CI), 4-Hexen-1-ol, 5-methyl-2-(1-methylethenyl)-, acetate (9CI), (±)-Lavandulol acetate, (±)-Lavandulyl acetate, 3-Acetoxymethyl-2,6-dimethyl-1,5-heptadiene, 5-Methyl-2-(1-methylethenyl)-4-hexen-1-yl acetate, 5-Methyl-2-(1-methylethenyl)-4-hexylene-1-ol acetate
IUPAC Name (5-methyl-2-prop-1-en-2-ylhex-4-enyl) acetate
SMILES CC(=CCC(COC(=O)C)C(=C)C)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H20O2/c1-9(2)6-7-12(10(3)4)8-14-11(5)13/h6,12H,3,7-8H2,1-2,4-5H3
Formula Weight 196.1463
Chemical Name or Material Lavandulyl Acetate