Learn More
SPEX CertiPrep™ Large Freezer Mill
Brand:  SPEX CertiPrep™ 6870D-230
Product Code. 15360472
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Fully programmable freezer mill that can store up to 10 user-defined methods for simple, fast and error free recall. A full color, detachable control screen is intuitive to use for quick and easy set-up and operation. A truly cryogenic mill that cools samples in liquid nitrogen and then pulverises them by magnetically shuttling a steel impactor between two steel end plugs within a sealed vial. The dual chamber allows for high throughput applications. Suitable for samples up to 200 grams (100 grams per chamber). This mill is supplied with a choice of one included accessory pack: if using large vials only, specify pack 6870L; if running small vials specify 6870S; if using midsize vials select, 6870M. If running more than one size of vial, more than one of these accessory packs will be needed, so order them as required.
  • Direct immersion in liquid nitrogen for optimum performance
  • Insulated case, nitrogen sensor and safety interlock ensures safe operation
  • Intuitive touch screen for setting grinding cycles, time, impact rate and cooling times
  • Sealed vials are suitable for volatile, temperature-sensitive and hazardous samples, and help eliminate cross-contamination
  • Pre-cooling chamber allows sample chilling while active chambers are running, helping to maximise productivity
  • Magnet-driven impactor is only moving part so practically maintenance-free
  • Each chamber takes large grinding vials for samples up to 100g, or four small grinding vials (6807 multivial adaptor needed)
  • Additionally each chamber can accommodate four microvial sets, meaning that up to 24 samples can be ground simultaneously
  • Grinding vials are available in a variety of materials inc polycarbonate, stainless steel and Cr-free, suitable for many applications
  • Supplied with 6804 extractor/vial opener and large vial rack
  • 6820 liquid nitrogen autofill supplied as standard Applications include: DNA/RNA extractions, forensics, materials testing, pharmaceutical analysis, drug testing, RoHS/WEEE and volatiles extraction. Typical samples include all types of plant and animal tissues, plastics and polymers, yeast, pharmaceuticals, food products, electronic components, textiles, hair, teeth and bones.