Learn More
MedChemExpress Larazotide acetate
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Purity | 98.0% |
| CAS | 881851-50-9 |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 785.89 g/mol |
| Product Type | Peptides |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 16.67 mg/mL (21.21 mM∣Need ultrasonic) ∣DMSO : 3.2 mg/mL (4.07 mM∣Need ultrasonic and warming) |