Learn More
Thermo Scientific™ LanthaScreen™ TR-FRET PPAR gamma Competitive Binding Assay Kit, rabbit
Description
This kit contains Rabbit Tb-Anti-GST antibody; other kit components are the same as kit PV4894:
The LanthaScreen™ TR-FRET PPAR gamma Competitive Binding Assay provides a sensitive and robust method for high-throughput screening (HTS) of ligands for peroxisome proliferator-activated receptor-gamma (PPAR gamma). The kit uses a terbium-labeled anti-GST antibody, a fluorescent small-molecule pan-PPAR ligand (Fluormone™ Pan-PPAR Green), and human PPAR gamma ligand-binding domain (LBD) that is tagged with glutathione S-transferase (GST) in a homogeneous mix-and-read assay format.
To assay:
When running the LanthaScreen™ TR-FRET PPAR gamma Competitive Binding Assay, Fluormone™ Pan-PPAR Green is added to ligand test compounds followed by addition of a mixture of the PPAR gamma-LBD and terbium anti-GST antibody. When the Fluormone™ Pan-PPAR Green is bound to PPAR gamma, energy transfer from the terbium-labeled antibody to the tracer occurs, and a high TR-FRET ratio is observed. Competitive ligand binding to PPAR gamma is detected by a test compound's ability to displace the tracer, which results in a loss of FRET between the antibody and the tracer. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the IC50 from a dose response curve of the compound (Figure 1). This type of binding assay is analogous to radioligand-based assays, except that it eliminates handling of radioactivity and enables a homogeneous, 'addition-only' format.
Specifications
Specifications
| Color | Green |
| Conjugate | Tb (Terbium) |
| Description | LanthaScreen™ TR-FRET PPAR Gamma Competitive Binding Assay Kit, 400 x 40μL Assays, Green, 384-Well Plate Format, Tb (Terbium) Dye Type, Fluorescent Detection Method, TR-FRET Technique, For Pharma & Biopharma, Target & Lead Identification |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Microplate Reader |
| Includes | Tb-anti-GST antibody (rabbit), PPAR gamma-LBD, GST, fluormone Pan-PPAR green, TR-FRET PPAR assay buffer, 1M DTT |
| Quantity | 400 x 40 μL assays |
| Regulatory Status | For research use only (RUO) |
| Packaging | 384-well Plate |
| Content And Storage | • Tb-anti-GST Antibody (Rabbit) (store at -20°C) • PPAR gamma-LBD, GST (-80°C) • Fluormone Pan-PPAR Green (-20°C) • TR-FRET PPAR Assay Buffer (4°C) • 1M DTT (-20°C or -80°C) |
| Show More |