missing translation for 'onlineSavingsMsg'
Learn More

LanthaScreen™ Eu-anti-GST Antibody

Product Code. p-7076004
Change view
Click to view available options
Quantity:
1 mg
25 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10645495 25 μg Each
10645685 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 10645495 Supplier Invitrogen Supplier No. PV5594
Add to basket
Add to basket
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

LanthaScreen™ Eu-anti-GST Antibody

LanthaScreen™ europium-labeled anti-GST goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GST
Applications Kinase Assay
Concentration 0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml
Conjugate Europium
Molecular Weight of Antigen 150kDa
Quantity 25 μg
Regulatory Status RUO
Research Discipline Kinase Biology
Primary or Secondary Primary
Packaging 1 Each
Content And Storage Europium-labeled anti-GST antibody in HEPES buffered saline. Store at -20°C.
Antibody Molecular Weight 150 kDa
Assay Entry Biochemical binding/kinetic
Excitation/Emission 340⁄665
For Use With (Equipment) Microplate Reader
Shipping Condition Dry Ice
Target Entry EPHA5, PDGFRA T674I, PDGFRA V561D, RET Y791F, SYK, TYRO3, MAP4K5, PRKX, ROCK2, IKBKE, PAK4, MAP3K2, LRRK2 D1994A, GRK7, GSK3A, SNF1LK2, ACVR2A, BMPR1A, RAF1 (cRAF) Y340D Y341D, MAP3K10, MAP3K9, JAK2 JH1 JH2, AMPK (A2/B1/G1), PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, MELK, EGFR, EPHB2, MERTK, RET V804L, ROS1, TEK, TTK, SGK, IKBKB, TAOK3, TYK2, IRAK1, ACVR1B, MYLK2, SRPK2, STK4, WNK2, FLT1, MUSK, LRRK2 R1441C, CSNK1G1, DYRK1A, DYRK3, RPS6KA6, DDR1, PTK2, PTK2B, RET, MINK1, GRK6, JAK3, LRRK2, AMPK (A1/B1/G1), CAMK4, PKN1, PRKD1, PRKG1, RPS6KA4, FLT4, INSR, EGFR L858R, ERBB4, TNK2, MAPK14, FRAP1, MAP3K3, EPHA, HCK, TAOK2, MAP3K11, CDK5, MAP2K3, TGFBR1, NLK, MAPK10, CSNK1G3, DMPK, DYRK1B, ACVR1, TGFBR2, EPHA1, EPHA8, EPHB3, MST1R, PDGFRA D842V, MAP4K4, mTOR, LRRK2 G2019S, CAMKK2, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, MARK4, RPS6KB1, EGFR T790M, EPHA7, EPHB1, PHKG1, ROCK1, BRAF V599E, JAK2, MARK1, PHKG2, MLCK, SLK, MKNK2, MYLK, FRK, ITK, BRAF, LRRK2 I2020T, BRSK1, CLK2, PIK3CD/PIK3R1, ACVR2B, MAP3K14, GRK5, GRK4, STK3, DDR2, SGKL, STK25, CLK1, CLK4, MARK2, STK17A, EIF2AK2, MAP3K5, EGFR T790M L858R, LIMK2
Target Molecule GST
Technique TR-FRET
Product Line LanthaScreen
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.