missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21997-1-AP-20UL
This item is not returnable.
View return policy
Description
Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Lamp1, also known as lysosome associated membrane protein 1, is a heavily glycosylated membrane protein of the lysosome. It contains a putative signal peptide, 18 sites for N-linked glycosylation, a single membrane-spanning segment and a short (11 amino acid) cytosolic tail.Specifications
| LAMP1 | |
| Polyclonal | |
| Unconjugated | |
| LAMP1 | |
| CD107a, LAMP 1, LAMP1, LAMPA, LGP120 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3916 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11279 | |
| LAMP1 | |
| LAMP1 Fusion Protein Ag17367 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction