Learn More
Laminin alpha 1 Rat anti-Mouse, Clone: AL-4, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems MAB4656-SP
Encompass
Product Code. 15737044
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Laminin alpha 1 Monoclonal specifically detects Laminin alpha 1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.