missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester Warning Signs

Product Code. 10132704
Click to view available options
Width (Metric):
50 mm
100 mm
200 mm
Displayed Text:
CAUTION Hot Surface Pictogram
Corrosive Batteries Warning Pictogram
Radioactive substances 7C III
Warning of Combustable Substances
Warning of Corrosive Substances
Warning of Gas Cylinders
Warning of Hazardous Substances
Warning of Oxidizing Substances
Warning of a magnetic field
Warning of suspended load
Unit Size:
1 roll
Each
This item is not returnable. View return policy

Product Code. 10132704

Brand: Brady™ 250243

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Material Polyester
Quantity 3/Pk.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 100 mm
Thickness 0.024 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Displayed Text Warning of Oxidizing Substances
Product Type Warning Sign
Color Black on Yellow
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.