Learn More
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs
Product Code. 11750204
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Quantity:
1 Ea.
250/Pk.
Unit Size:
1 roll
Each
Product Code. 11750204
Brand: Brady™ 223556
Only null left
Add to basket
Add to basket
Specifications
Alternate Part Number ADR 1.5-200*200-B7541
Finish Gloss
Material Laminated Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Height (Metric) 200 mm
Thickness 0.099 mm
Width (Metric) 200.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Diamond
Displayed Text 1.5 1
Color Black on Orange