Learn More
Bochem™ Ladle Scoop
Product Code. 10120161
Click to view available options
Capacity (Metric):
200 mL
220 mL
Unit Size:
Each
Product Code. 10120161
Brand: Bochem™ 8741
Please to purchase this item. Need a web account? Register with us today!
Manufactured from stainless steel. Bochem™ Ladle Scoop has a flat handle.
  • Ladle scoop
  • Flat handle
  • Stainless steel
Specifications
Shape Flat
Capacity (Metric) 200 mL
Certifications/Compliance AISI 304
Product Type Ladle Scoop
Material Stainless Steel (18/10)
Handle Type Flat
Length (Metric) 315 mm