Learn More
LABPLAS LABRACK
Product Code. 17930225
Click to view available options
Holds:
Bags
Unit Size:
Each
Product Code. 17930225
Brand: LABPLAS SA47012
Only null left
Add to basket
Add to basket
Designed for the safe transport or storage of sample bags.
  • Designed for the safe transport or storage of sample bags
  • Stackable, autoclavable and can be used in sterile and refrigerated environments
  • Holds 10 - 400 series bags
  • Material: Stainless steel
Specifications
Product Type Bag Rack
Autoclavable Autoclavable
Holds Bags
Material Stainless Steel
Shape Rectangular
Stackable Yes
Dimensions (L x W) 197 x 419 mm