Learn More
LABPLAS LABRACK
Click to view available options
Holds:
Bags
Unit Size:
Each
Description
- Designed for the safe transport or storage of sample bags
- Stackable, autoclavable and can be used in sterile and refrigerated environments
- Holds 10 - 400 series bags
- Material: Stainless steel
Specifications
Specifications
| Product Type | Bag Rack |
| Autoclavable | Autoclavable |
| Holds | Bags |
| Material | Stainless Steel |
| Shape | Rectangular |
| Stackable | Yes |
| Dimensions (L x W) | 197 x 419 mm |