Learn More
behr Labor-Technik™ Laboratory Peristaltic Pump
Product Code. 10014901
Click to view available options
Flow Rate:
0.4 to 2 L/hr
1 to 4 L/hr
15 to 60 L/hr
Unit Size:
Each
Product Code. 10014901
Brand: behr Labor-Technik™ B00210220
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compact laboratory peristaltic pump with continuously adjustable conveying capacity, pump rotor with two compression and four guide rollers. Changing hose is easy, no tools or other aids are required. AC motors 220V∼, 50 Hz.
  • The speed of the precision DC motor can be adjusted infinitely
    • The rotational direction of the pump rotor can be reversed
Specifications
Flow Rate 15 to 60 L/hr