Learn More
Knick Elektronische™ 703 Laboratory Conductivity Meter
Description
- Measuring ranges from 0.000 μS/cm to 2000 mS/cm
- 1.000 μS/cm to >1000 mS/cm with one sensor
- Records for QM documentation according to ISO 9000 and GLP
- Calibrated analog recorder output, galvanically isolated
- Sensoface: monitoring of the sensor and equipment
- Automatic calibration with standard solutions
- EMC according to NAMUR
- RS-232 interface for computer and printer
- Simultaneous display of two measurement values
- Self-contained clock
- Leak-proof membrane keyboard
- Robust housing
- Protection category IP 54
- 3-year warranty
Specifications
Specifications
| Includes | RS-232 interface for computer and printer, Self-contained clock, Leak-proof membrane keyboard, Robust housing |
| Calibration | Automatic calibration with standard solutions |
| Range (Conductivity) | Measuring ranges from 0.000 μS/cm to 2000 mS/cm, 1.000 μS/cm to >1000 mS/cm with one sensor |
| Model | 703 Laboratory Conductivity Meter |
| Keypad | Leak-proof membrane keyboard |
| Interface | RS-232 |
| Type | Conductivity Meter |