Learn More
SHP Steriltechnik AG Laboklav 25 Insert, 4 Levels
Description
- Insert with 4 Levels for Laboklav 25
- Stainless steel
- WxHxD (mm): 190 x 426,5 x 180
Specifications
Specifications
| Capacity (Metric) | Not Applicable |
| For Use With (Equipment) | Laboklav 25 |
| Model | Insert, 4 Levels |
| Product Type | Shelf |
| Sterility | Non-sterile |