Learn More
Sampling Systems™ Label Dispenser for 95mm Wide Labels
Product Code. 12744758 Shop All Sampling Systems Products
Click to view available options
:
ProductTypeSpecs: Label Dispenser
Product Code. 12744758
Brand: Sampling Systems™ 8220A07
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sampling Systems™ Label Dispenser for 95mm Wide Labels is an easy way to store and dispense Ultra Adhesive Quality Control Labels.
  • Holds one roll of labels
  • Easy to use
Specifications
Product Type Label Dispenser
For Use With (Equipment) 95 x 95 mm wide labels