Learn More
Labconco™ HEPA Filter
For Purifier Class I and HEPA-Filtered Enclosures and XPert and RXPert Enclosures.
Brand:  Labconco™ 3707902
Dettagli aggiuntivi : Weight : 71.70000kg
Codice prodotto. 15868946
  • 835.00 € / Each
Estimated Shipment: 28-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Descrizione
Descrizione
Supply HEPA filter for 4' Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert & RXPert Filtered Balance Systems.
Specifica
Specifica
99.99%
4 ft. Purifier™ class I safety, HEPA filtered, vertical clean bench and PCR enclosure
71.7 Kg
101.6 to 127 cm
42 in.
3.3 in.
HEPA
158 lb.
40 to 50 in.
18 in.
Particulate
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Product Certifications
Offerte speciali
Offerte speciali