Learn More
Vascular Endothelial Growth Factor (VEGF) Ab-7 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: VG1
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant VEGF protein
Molecular Weight: 19-22kDa (reduced)
Positive Control: VEGF165 recombinant protein, Angiosarcoma Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF. Recommended for: Note: USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Vascular Endothelial Growth Factor (VEGF) Ab-7 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | VG1 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant VEGF protein |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Research Discipline | Angiogenesis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.