Learn More
Description
Host Species: Mouse
Clone: A4.1
Isotype: IgM
Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.
Epitope: N-terminal half of the central stalk-like region of TSP, which is recovered as a 50kDa fragment after chymotryptic digestion of TSP
Immunogen: Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets
Molecular Weight: ∽450kDa (nonreduced)
Positive Control: HUVEC cells
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting (Nonreduced only)
- Immunofluorescence
- Blocks Binding of TSP-1 to TSP-Rec./CD36
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Thrombospondin (TSP) Ab-1 |
| Applications | Flow Cytometry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | A4.1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.