missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin (TSP) Ab-1 Mouse Monoclonal Antibody, Epredia™

Product Code. 12641377 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
Unit Size:
100µL
1mL
200µL
500µL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration unitSize
12641377 100 μL 200μg/mL 100µL
12651377 500 μL 200μg/mL 500µL
12631377 1 mL 200μg/mL 1mL
12661377 100 μL 1mg/mL 100µL
12671377 200 μL 1mg/mL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12641377 Supplier Epredia Supplier No. MS418P0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide accurate, reproducible results with the Epredia™ Thrombospondin (TSP) Ab-1, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A4.1

Isotype: IgM

Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.

Epitope: N-terminal half of the central stalk-like region of TSP, which is recovered as a 50kDa fragment after chymotryptic digestion of TSP

Immunogen: Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced)

Positive Control: HUVEC cells

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Nonreduced only)
  • Immunofluorescence
  • Blocks Binding of TSP-1 to TSP-Rec./CD36
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin (TSP) Ab-1
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone A4.1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Rat
Form Concentrated
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.