Learn More
Epredia™ Lab Vision™ PARP (Poly ADP-Ribose Polymerase) Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide for the N-terminal region of human PARP
Molecular Weight: 116kDa
Positive Control: Raji cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | PARP (Poly ADP-Ribose Polymerase) Ab-2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide for the N-terminal region of human PARP |
| Purification Method | Purified w/o BSA and Azide |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.