Learn More
Description
Host Species: Mouse
Clone: CA-4001 or CA719E3C
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: N-terminal (APSPIIKFPGD-VAPKTDK)
Immunogen: N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV
Molecular Weight: 72kDa (proform)
Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells
Cellular Localization: Cytoplasmic
Recommended for:
- Western Blotting
- Immunoprecipitation (Not suitable)
- Inhibition of MMP-2 Activity
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-2 (72kDa Collagenase IV) Ab-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CA-4001 or CA719E3C |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.