Learn More
Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X)
Product Code. 12668286 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12668286 500mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12668286 Supplier Epredia™ Supplier No. AP9004500
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Improve the reactivity of many antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ EDTA Buffer for Heat-Induced Epitope Retrieval (10X).
  • 10mM EDTA buffer, pH 8.0
  • Dilute 10-fold with distilled water before use
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives. For periods longer than 3 months, store at 2-8°C
TRUSTED_SUSTAINABILITY
Spezifikation
Content And Storage Room temperature
Dilution 10X
Product Type EDTA
Quantity 500mL
For Use With (Application) Immunohistochemistry
pH 8