missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1065-P0

 View more versions of this product

Product Code. 12658796

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Mouse

Clone: CG04 + CG05

Isotype: IgG2a/κ + IgG1/κ

Species Reactivity: Human. Others not known

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chorionic Gonadotropin, Human/hCG Ab-5
Monoclonal
200μg/mL
Mouse
100 μL
Steroids Hormones and Growth Factors
Human
IgG1 κ
Immunohistochemistry (Paraffin)
CG04 + CG05
Unconjugated
Purified Beta-subunit of HCG
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.