Learn More
WTW™ Lab Station for pHotoFlex™ Series and Turb 430 Models
Click to view available options
Type:
Lab Station
Unit Size:
Each
Specifications
Specifications
| Type | Lab Station |
| For Use With (Application) | Used as bar code reader, easy lab use with software package for GLP-compliant data handling, loading function |
| For Use With (Equipment) | For pHotoFlex™ series and Turb™ 430 models |
| Includes | PC-Software LSdata, rechargeable battery set, universal plug, RS-232 interface, cable |