Learn More
WTW™ Lab Station for pHotoFlex™ Series and Turb 430 Models
GREENER_CHOICE
Product Code. 10601233
Click to view available options
Type:
Lab Station
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 10601233
Brand: WTW™ 251301
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Specifications
Type Lab Station
For Use With (Application) Used as bar code reader, easy lab use with software package for GLP-compliant data handling, loading function
For Use With (Equipment) For pHotoFlex™ series and Turb™ 430 models
Includes PC-Software LSdata, rechargeable battery set, universal plug, RS-232 interface, cable