Learn More
VACUUBRAND™ Lab Scaffold Mounting Assembly Kit
Product Code. 15928632
Change view
Click to view available options
For Use With:
CVC 3000, DCP 3000
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
15928632 CVC 3000, DCP 3000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15928632 Supplier VACUUBRAND™ Supplier No. 22614163
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For mounting CVC 3000 and DCP 3000 in lab scaffolding
TRUSTED_SUSTAINABILITY
Specifications
For Use With CVC 3000, DCP 3000
Quantity 1 Ea.
Type Assembly Kit