Learn More
L-Tryptophan-2',4',5',6',7'-d5 (indole-d5), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Tryptophan-d5 |
| CAS | 62595-11-3 |
| Molecular Formula | C11 2H5 H7 N2 O2 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13 H,5,12H2,(H,14,15)/t9-/m0/s1/i1D,2 D,3 D,4 D,6 D |
| SMILES | [2 H]c1[nH]c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1C[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-3-(2,4,5,6,7-pentadeuterio-1 H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 209.26 |
| Formula Weight | 209.1213 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |