Learn More
L-Serine-d7, CDN
Product Code. 30311834
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311834 0.1 g 0.1g
30325601 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311834 Supplier CDN Supplier No. CDND64660.1G
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Serine-d7
CAS 935275-35-7
Molecular Formula C3 D7 N O3
Quantity 0.1 g
Synonym L-Serine-N,N,O,1,2,3,3-d7 (ACI), L-Serine-d7, L-Serine-D7, (2 S)-2-(Amino-d2)-3-(hydroxy-d)propanoic-2,3,3-d3 acid-d
InChI Formula InChI=1 S/C3H7NO3/c4-2(1-5)3(6)7/h2,5 H,1,4H2,(H,6,7)/t2-/m0/s1/i1D2,2 D,5 D/hD3
SMILES [2 H]OC(=O)[C@@]([2 H])(N([2 H])[2 H])C([2 H])([2 H])O[2 H]
IUPAC Name deuterio (2 S)-2,3,3-trideuterio-3-deuteriooxy-2-(dideuterioamino)propanoate
Molecular Weight (g/mol) 112.1357
Formula Weight 112.0865 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity