Learn More
L-Serine -13C3,15N, TRC
Product Code. 30011405
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011405 2.5 mg 2.5mg
30004212 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30011405 Supplier Toronto Research Chemicals Supplier No. TRCS2709972.5MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 202407-34-9
Molecular Formula 13C3H715NO3
Molecular Weight (g/mol) 109.064
Synonym L-Serine-1,2,3-13C3-15N (ACI), L-Serine-13C3-15N (9CI), L-Serine-13C3,15N, (2S)-2-(Amino-15N)-3-hydroxypropanoic-1,2,3-13C3 acid
IUPAC Name (2S)-2-(^{15}N)azanyl-3-hydroxy-(1,2,3-^{13}C_{3})propanoic acid
SMILES [15NH2][13C@@H]([13CH2]O)[13C](=O)O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i1+1,2+1,3+1,4+1
Formula Weight 109.0497
Chemical Name or Material L-Serine-(13C3,15N)