Learn More
L-Pipecolic acid, MedChemExpress
Product Code. 16499356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16499356 Supplier Medchem Express Supplier No. HYW012734
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress L-Pipecolic acid (H-HoPro-OH) is a breakdown product of lysine, accumulates in body fluids of infants with generalized genetic peroxisomal disorders, such as Zellweger syndrome, neonatal adrenoleukodystrophy.
L-Pipecolic acid (H-HoPro-OH) is a breakdown product of lysine, accumulates in body fluids of infants with generalized genetic peroxisomal disorders, such as Zellweger syndrome, neonatal adrenoleukodystrophy.
Specifications
Chemical Name or Material L-Pipecolic acid
CAS 3105-95-1
Color White
Purity Grade Notes Research
Molecular Formula C6H11NO2
Quantity 100 mg
Synonym H-HoPro-OH
Solubility Information H2O : 100 mg/mL (774.23 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (7.74 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C([C@H]1NCCCC1)O
Molecular Weight (g/mol) 129.16
Formula Weight 129.16
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism