missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals

Product Code. p-7041730
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15462889

Brand: Thermo Scientific Alfa Aesar J60043.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes
  • These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2577-40-4
Molecular Formula C18H20N2O3
Molecular Weight (g/mol) 312.369
MDL Number MFCD00063154
InChI Key GKZIWHRNKRBEOH-HOTGVXAUSA-N
Synonym h-phe-phe-oh, l-phe-l-phe, s-2-s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, chembl55868, phe-phe, 2s-2-2s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, l-phenylalanyl-l-phenylalanine, phenylalanyl-phenylalanine, 2s-2-2s-2-amino-3-phenylpropanoyl amino-3-phenylpropanoic acid
PubChem CID 6993090
ChEBI CHEBI:72723
IUPAC Name (2S)-2-[[(2S)-2-amino-3-phenylpropanoyl]amino]-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)NC(CC2=CC=CC=C2)C(=O)O)N

Specifications

Color White
Solubility Information Soluble at 50mg/ml in 80% acetic acid
Quantity 1 g
Formula Weight 312.37
Physical Form Powder
Chemical Name or Material L-Phenylalanyl-L-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.