Learn More
L-Phenylalanine-13C9 (9CI), TRC
Specifications
Specifications
| Chemical Name or Material | L-Phenylalanine-13C9 |
| CAS | 439685-11-7 |
| Molecular Formula | 13C9 H11 N O2 |
| Quantity | 1 mg |
| Synonym | L-Phenylalanine-carboxy,α,β,1,2,3,4,5,6-13C9, L-Phenylalanine-13C9 (9CI), L-13C9-Phenylalanine, L-Phenylalanine-13C9, L-[13C9]Phenylalanine, (2S)-2-Amino-3-(phenyl-13C6)propanoic-1,2,3-13C3 acid, (S)-Phenylalanine-13C9 |
| InChI Formula | InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1 |
| SMILES | N[13C@@H]([13CH2][13c]1[13cH][13cH][13cH][13cH][13cH]1)[13C](=O)O |
| IUPAC Name | (2S)-2-amino-3-((1,2,3,4,5,6-^{13}C_{6})cyclohexatrienyl)(1,2,3-^{13}C_{3})propanoic acid |
| Molecular Weight (g/mol) | 174.123 |
| Formula Weight | 174.109 |