Learn More
L-Phenylalanine-13C6, TRC
Product Code. 30008990
Change view
Click to view available options
Quantity:
2.5 mg
100 mg
250 mg
Unit Size:
100mg
2.5mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008990 2.5 mg 2.5mg
30006631 100 mg 100mg
30006473 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008990 Supplier Toronto Research Chemicals Supplier No. TRCP3194172.5MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 180268-82-0
Molecular Formula C313C6H11NO2
Molecular Weight (g/mol) 171.15
Synonym (2S)-2-Amino-3-phenylpropanoic Acid-13C6, (S)-(-)-Phenylalanine-13C6, (S)-alpha-Amino-beta-phenylpropionic Acid-13C6, (S)-alpha-Aminohydrocinnamic Acid-13C6, (S)-alpha-Amino-benzenepropanoic Acid-13C6;
SMILES N[C@H](C(O)=O)C[13C]1=[13C]([H])[13C]([H])=[13C]([H])[13C]([H])=[13C]1[H]
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)
Formula Weight 171.0991
Chemical Name or Material L-Phenylalanine-13C6