Learn More
L-O-Phosphoserine-13C315N, TRC
Product Code. 30426214
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426214 0.5 mg 0.5mg
30410347 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30426214 Supplier Toronto Research Chemicals Supplier No. TRCP3612020.5MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2734706-69-3
Molecular Formula 13C3 H8 15N O6 P
Molecular Weight (g/mol) 189.04
Synonym Dihydrogen phosphate L-Serine-13C315N, L-Serine Phosphate-13C315N, L-Serine Dihydrogen Phosphate-13C315N, 3-O-Phosphoserine-13C315N, Dexfosfoserine-13C315N, Fosforina-13C315N, L-3-Phosphoserine-13C315N, L-O-Phosphoserine-13C315N, L-O-Serine phosphate-13C315N, L-Phosphoserine-13C315N, L-Serinephosphoric Acid-13C315N, L-Seryl Phosphate-13C315N, O-Phospho-L-serine-13C315N, O-Phosphoryl-L-serine-13C315N, O-Phosphorylserine-13C315N, Phospho-L-serine-13C315N, Serine O-phosphate-13C315N, Serine Dihydrogen Phosphate-13C315N, Seriphos-13C315N
IUPAC Name (2S)-2-(^{15}N)azanyl-3-phosphonooxy-(1,2,3-^{13}C_{3})propanoic acid
SMILES OP(O[13CH2][13C@H]([15NH2])[13C](O)=O)(O)=O
Specifications
Quantity 0.5 mg
InChI Formula InChI=1S/C3H8NO6P/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H2,7,8,9)/t2-/m0/s1/i1+1,2+1,3+1,4+1
Percent Purity >95%
Chemical Name or Material L-O-Phosphoserine-13C315N
Grade HPLC